missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK3 Antibody (SJB3-36), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04261PE
This item is not returnable.
View return policy
Description
AK3 Monoclonal antibody specifically detects AK3 in Human samples. It is validated for Flow Cytometry
Specifications
| AK3 | |
| Monoclonal | |
| PE | |
| Adenylate kinase 3 alpha-like 1, adenylate kinase 3 like 1, adenylate kinase 3adenylate kinase 6, AK 3, AK6, AKL3L, AKL3L1, EC 2.7.4, EC 2.7.4.10, FIX, GTP:AMP phosphotransferase, mitochondrial | |
| Recombinant human AK3 (adenylaste kinase isozyme 3) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 50808 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SJB3-36 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction