missing translation for 'onlineSavingsMsg'
Learn More

AK3 Monoclonal Antibody (OTI6D6), TrueMAB™, OriGene

Product Code. p-200009659
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17595291 30 μL 30µL
17575291 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17595291 Supplier Origene Technologies Supplier No. TA501833S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AK3 Monoclonal antibody specifically detects AK3 in Human samples. It is validated for Western Blot

Specifications

Antigen AK3
Applications Western Blot
Classification Monoclonal
Clone OTI6D6
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Gstt2
Gene Accession No. P0CG29
Gene Alias AA407498; adenylate kinase 3; adenylate kinase 3 {ECO:0000255; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255; adenylate kinase 6, adenylate kinase 3 like 1; AI506714; AK 3; AK 3 {ECO:0000255; AK3; AK-3; Ak3l; AK3L1; AK4; AK6; AKL3L; AKL3L1; FIX; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial; HAMAP-Rule:MF_03169}
Host Species Mouse
Immunogen Full length human recombinant protein of human GSTT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2953
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.