Learn More
AK3L1 Antibody (010), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90093PCP
Product Code. 18699571
  • € 607.00 / 0.10mL
Description
AK3L1 Monoclonal antibody specifically detects AK3L1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
0.1 mL
Store at 4C in the dark.