missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK3L1 Antibody (014), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90094R
This item is not returnable.
View return policy
Description
AK3L1 Monoclonal specifically detects AK3L1 in Human samples. It is validated for ELISA.
Specifications
| AK3L1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 14 | |
| ELISA | |
| adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human AK3L1 (P27144-1; Ala2-Tyr223). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 205 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction