missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK5 Antibody (OTI2F9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71813PCP
This item is not returnable.
View return policy
Description
AK5 Monoclonal specifically detects AK5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| AK5 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| AK5 | |
| Full length recombinant protein of human AK5 (NP_777283) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI2F9 | |
| Flow Cytometry | |
| adenylate kinase 5, adenylate kinase 6, adenylate kinase isoenzyme 5, AK 5, AK6, ATP-AMP transphosphorylase 5, EC 2.7.4.3, MGC33326 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 26289.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction