missing translation for 'onlineSavingsMsg'
Learn More

AKAP10 Polyclonal Antibody, Invitrogen™

Product Code. 15535451
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535451 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535451 Supplier Invitrogen Supplier No. PA538529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKAP 10 (A kinase anchor protein 10), also known as PRKA10 or D-AKAP2 (dual-specific A kinase-anchoring protein 2), is a 662 amino acid mitochondrial membrane protein that belongs to the AKAP family. AKAP 10 is a dual specificity protein that binds to both type I and type II regulatory subunits of PKA and anchors them to the plasma membrane or the mitochondria. When anchored to the mitochondria, PKA can phosphorylate and, thus, inactivate the proapoptotic protein Bad. This suggests that AKAP 10 indirectly regulates Bad-induced apoptosis by mediating the mitochondrial attachment of PKA. Additionally, AKAP 10 may facilitate G-protein coupled signal transduction and could act as an adaptor in the assembly of multiprotein complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP10
Gene Accession No. O43572, O88845
Gene Alias A-kinase anchor protein 10, mitochondrial, AKAP-10, Dual specificity A kinase-anchoring protein 2, D-AKAP-2, Protein kinase A-anchoring protein 10, PRKA10, AKAP10
Gene Symbols AKAP10
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human AKAP10
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11216, 360540, 56697
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.