missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118922
This item is not returnable.
View return policy
Description
AKAP11 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed at high levels throughout spermatogenesis and in mature sperm. It binds the RI and RII subunits of PKA in testis. It may serve a function in cell cycle control of both somatic cells and germ cells in addition to its putative role in spermatogenesis and sperm function. [provided by RefSeq, Jul 2008].
Specifications
| AKAP11 | |
| Polyclonal | |
| Unconjugated | |
| AKAP11 | |
| A kinase (PRKA) anchor protein 11; a kinase anchor protein 220 kDa; AKAP 220; AKAP11; AKAP-11; AKAP220; A-kinase anchor protein 11; A-kinase anchor protein 220 kDa; A-kinase anchoring protein 11; A-kinase anchoring protein, 220kDa; KIAA0629; PPP1R44; PRKA11; protein kinase A anchoring protein 11; protein kinase A-anchoring protein 11; protein phosphatase 1, regulatory subunit 44 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11215 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9UKA4 | |
| AKAP11 | |
| Recombinant protein A Kinase Anchor Protein 11. The antigen corresponds to amino acid range 1650-1901 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur