missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | AKAP3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
AKAP3 Polyclonal antibody specifically detects AKAP3 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| AKAP3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 10566 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| A kinase (PRKA) anchor protein 3, AKAP110AKAP 110, AKAP-3, A-kinase anchor protein 110 kDa, Cancer/testis antigen 82, CT82A-kinase anchor protein 3, Fibrous sheath protein of 95 kDa, fibrousheathin 1, Fibrousheathin I, Fibrousheathin-1, FSP95A-kinase anchor protein, 110kDa, PRKA3, protein kinase A anchoring protein 3, Protein kinase A-anchoring protein 3, SOB1fibrous sheath protein, 95kDa, Sperm oocyte-binding protein, sperm oocyte-binding protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human AKAP3 (NP_006413.3). MSEKVDWLQSQNGVCKVDVYSPGDNQAQDWKMDTSTDPVRVLSWLRRDLEKSTAEFQDVRFKPGESFGGETSNSGDPHKGFSVDYYNTTTKGTPERLHFEMTHKEIPCQGPRAQLGNGSSVDEVSFYANRLTNLVIAMARKEINEKIDGSENKCVYQSLYMGNEPTPTKSLSKIASELVNETVSACSRNAAPDKAPGSGDRVSGSSQSPPNLKYKSTLKIKESTKERQGPDDKPPSKKSFFYKEVFESRNGDYAREGGRFFPRERKRFRGQERPDDFTASVSEGIMTYANSVVSDMMVSI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title