missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP7 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97418FR
This item is not returnable.
View return policy
Description
AKAP7 Polyclonal specifically detects AKAP7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKAP7 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| A kinase (PRKA) anchor protein 7, AKAP 18, AKAP15, AKAP18A-kinase anchor protein 7 isoform alpha, AKAP-7 isoform gamma, AKAP-7 isoforms alpha and beta, A-kinase anchor protein 18 kDa, A-kinase anchor protein 7, A-kinase anchor protein 7 isoform gamma, A-kinase anchor protein 7 isoforms alpha and beta, A-kinase anchor protein 9 kDa, A-kinase anchor protein, 18-kD, A-kinase anchoring protein 18, protein kinase A anchoring protein 7, Protein kinase A-anchoring protein 7 isoform gamma, Protein kinase A-anchoring protein 7 isoforms alpha/beta | |
| Produced in rabbits immunized with E. coli-derived Human AKAP7 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9465 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction