missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP7 Antibody (OTI6F7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71466H
This item is not returnable.
View return policy
Description
AKAP7 Monoclonal specifically detects AKAP7 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AKAP7 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| AKAP7 | |
| Full length recombinant protein of human AKAP7 (NP_057461) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI6F7 | |
| Western Blot, Immunohistochemistry | |
| A kinase (PRKA) anchor protein 7, AKAP 18, AKAP15, AKAP18A-kinase anchor protein 7 isoform alpha, AKAP-7 isoform gamma, AKAP-7 isoforms alpha and beta, A-kinase anchor protein 18 kDa, A-kinase anchor protein 7, A-kinase anchor protein 7 isoform gamma, A-kinase anchor protein 7 isoforms alpha and beta, A-kinase anchor protein 9 kDa, A-kinase anchor protein, 18-kD, A-kinase anchoring protein 18, protein kinase A anchoring protein 7, Protein kinase A-anchoring protein 7 isoform gamma, Protein kinase A-anchoring protein 7 isoforms alpha/beta | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 9465.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction