missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP7 Monoclonal Antibody (OTI2A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804913
This item is not returnable.
View return policy
Description
This gene encodes a member of the A-kinase anchoring protein family, a group of functionally related proteins that bind to a regulatory subunit of cAMP-dependent protein kinase A and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.Specifications
| AKAP7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43687 | |
| AKAP7 | |
| Full length human recombit protein of human AKAP7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2A7 | |
| Unconjugated | |
| AKAP7 | |
| AKAP15, AKAP18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9465 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction