missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552103
This item is not returnable.
View return policy
Description
Immunogen sequence: PEMEGDDNLG GEDKKETPEE VAADVLAEVI TAAVRAVDGE GAPAPESSGE PAEDEGPTDT AEAGSDPQAE QLLEEQVPCG TAHEKGVPKA RSEAAEAGNG AETMAAEAES AQTRVAPAPA AADAEVEQTD AESKDAVP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 58%.
The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 95, also known as AKAP 8, is a nuclear matrix protein predominantly expressed in liver, heart, pancreas, kidney and skeletal muscle. During mitosis, AKAP 95 is recruited to the chromosomes and plays an essential role in mitotic progression. Characteristic of its family, AKAP 95 participates in PKA signaling through an interaction with the RII regulatory subunit. In addition, AKAP 95 forms a complex with HA95 and HDAC3 and is required for the deacetylation of Histone H3 in mitosis.
Specifications
| AKAP8 | |
| Polyclonal | |
| Unconjugated | |
| AKAP8 | |
| 1200016A02Rik; A kinase (PRKA) anchor protein 8; A kinase anchor protein 8; AA673585; AKAP 8; AKAP 95; Akap8; AKAP-8; Akap95; AKAP-95; A-kinase anchor protein 8; A-kinase anchor protein 95 kDa; A-kinase anchor protein, 95kDa; A-kinase anchoring protein 8; AU015639; DKFZp586B1222 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43823 | |
| AKAP8 | |
| Recombinant protein corresponding to Human AKAP8. Recombinant protein control fragment (Product #RP-102489). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto