missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP9 Antibody (17G10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 200.00
Specifications
| Antigen | AKAP9 |
|---|---|
| Clone | 17G10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:100 |
| Applications | Western Blot |
Description
AKAP9 Monoclonal specifically detects AKAP9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| AKAP9 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 10142 | |
| Synthetic peptide corresponding to residues Q(31) F R Q R K A Q S D G Q S P S(45) of human AKAP450. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 17G10 | |
| Western Blot 2 μg/mL, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| A kinase (PRKA) anchor protein (yotiao) 9, AKAP 350, AKAP 450, AKAP350A-kinase anchor protein 450 kDa, AKAP450A-kinase anchor protein 350 kDa, AKAP-9, Centrosome- and Golgi-localized PKN-associated protein, CG-NAPAKAP 120-like protein, hgAKAP 350, HYPERION, KIAA0803A-kinase anchor protein 9, MU-RMS-40.16A, PRKA9AKAP9-BRAF fusion protein, Protein hyperion, protein kinase A anchoring protein 9, Protein kinase A-anchoring protein 9, Protein yotiao, YOTIAOkinase N-associated protein | |
| AKAP9 | |
| IgG | |
| Protein G purified | |
| AKAP450 (17G10) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title