missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP9 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-86994C
This item is not returnable.
View return policy
Description
AKAP9 Polyclonal antibody specifically detects AKAP9 in Human samples. It is validated for Western Blot
Specifications
| AKAP9 | |
| Polyclonal | |
| Western Blot | |
| A kinase (PRKA) anchor protein (yotiao) 9, AKAP 350, AKAP 450, AKAP350A-kinase anchor protein 450 kDa, AKAP450A-kinase anchor protein 350 kDa, AKAP-9, Centrosome- and Golgi-localized PKN-associated protein, CG-NAPAKAP 120-like protein, hgAKAP 350, HYPERION, KIAA0803A-kinase anchor protein 9, MU-RMS-40.16A, PRKA9AKAP9-BRAF fusion protein, Protein hyperion, protein kinase A anchoring protein 9, Protein kinase A-anchoring protein 9, Protein yotiao, YOTIAOkinase N-associated protein | |
| The immunogen recognized by this antibody maps to a region between residue 1075 and 1125 of human A kinase (PRKA) anchor protein 9 (centrosome- and golgi-localized PKN-associated protein) using the numbering given in entry NP_671700.1 (GeneID 10142). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 10142 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction