missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110913
This item is not returnable.
View return policy
Description
Immunogen sequence: KLLEAISETS SQLEHAKVTQ TELMRESFRQ KQEATESLKC QEELRERLHE ESRAREQLAV ELSKAEGVID GYADEKTLFE RQIQEKTDII DRLEQELLCA SNRLQELEAE QQQIQEEREL LSRQKEAMKA EAGPVEQQLL QETE Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 86%.
The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
Specifications
| AKAP9 | |
| Polyclonal | |
| Unconjugated | |
| AKAP9 | |
| 5730481H23Rik; A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; AKAP 120-like protein; AKAP 350; AKAP 450; AKAP350; AKAP450; AKAP9; AKAP-9; AKAP9-BRAF fusion protein; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchor protein 9; A-kinase anchoring protein 9; AW545847; C79026; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; CG-NAP; G1-448-15; HYPERION; KIAA0803; kinase N-associated protein; LQT11; mei2-5; mKIAA0803; MU-RMS-40.16A; PPP1R45; PRKA9; Protein hyperion; protein kinase A anchoring protein 9; protein kinase A-anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; repro12; YOTIAO | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10142 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99996 | |
| AKAP9 | |
| Recombinant Protein corresonding to Human AKAP9. Recombinant protein control fragment (Product #RP-102408). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction