missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKIRIN1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98595UV
This item is not returnable.
View return policy
Description
AKIRIN1 Polyclonal antibody specifically detects AKIRIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AKIRIN1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| AKIRIN1 akirin 1, C1orf108, RP11-781D11.2, STRF2 | |
| Produced in rabbits immunized with E. coli-derived Human AKIRIN1 fragment. | |
| 0.1 mL | |
| Innate Immunity | |
| 79647 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction