missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKNA Polyclonal Antibody
GREENER_CHOICE

Product Code. 15596942
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15596942 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15596942 Supplier Invitrogen™ Supplier No. PA545800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: AGGAVTGDPL GPPPADTLQC PLCGQVGSPP EADGPGSATS GAEKATTRRK Sequence homology: Human: 100%; Rabbit: 92%; Rat: 85%.

AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKNA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AKNA
Gene Accession No. Q7Z591
Gene Alias AKNA; AKNA transcript F2; AT-hook transcription factor; AT-hook transcription factor AKNA; AT-hook-containing transcription factor; KIAA1968; Microtubule organization protein AKNA
Gene Symbols AKNA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human AKNA (aa 1252-1301).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80709
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.