missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AKOS 022

Description
AKOS 022 is a voltage-dependent anion channel (VDAC; Kd = 154 μM) inhibitor, which reduces channel conductance. In HEK-293 cells, AKOS 022 inhibits selenium-induced VDAC oligomerization, cytochrome C release from mitochondria and apoptosis (IC50 values range from 3.3 to 3.6 μM). AKOS 022 also prevents glucotoxicity-mediated overexpression of VDAC1 in INS-1 cells.
Specifications
Specifications
| Chemical Name or Material | 1-(4-Chlorophenyl)-3-[4-[[4-(trifluoromethoxy)phenyl]amino]-1-piperidinyl]-2,5-pyrrolidinedione |
| CAS | 878983-38-1 |
| Quantity | 10 mg |
| Target | Ion Channel Modulators |
| Molecular Formula | C22H21ClF3N3O3 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?