Learn More
AKR1A1 Mouse anti-Human, Clone: OTI10E11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500744
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.Specifications
| AKR1A1 | |
| Monoclonal | |
| Unconjugated | |
| AKR1A1 | |
| ALDR1, ALR, ARM, DD3, HEL-S-6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10327 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI10E11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P14550 | |
| AKR1A1 | |
| Full length human recombit protein of human AKR1A1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.