missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Antibody (109), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89882H
This item is not returnable.
View return policy
Description
AKR1B1 Monoclonal specifically detects AKR1B1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| AKR1B1 | |
| Monoclonal | |
| HRP | |
| ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human AKR1B1 (P15121; Met1-Phe316). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 231 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 109 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction