missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Antibody (AKR1B1/7009R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14283JF669
This item is not returnable.
View return policy
Description
AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| AKR1B1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
| Recombinant human full-length AKR1B1 protein (Uniprot: P15121) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 231 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| AKR1B1/7009R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction