missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99771-100ul
This item is not returnable.
View return policy
Description
AKR1B1 Polyclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| AKR1B1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
| Produced in rabbits immunized with purified, recombinant Human AKR1B1 (Uniprot#: P15121; Met1-Phe316). | |
| 100 μg | |
| Stem Cell Markers | |
| 231 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction