missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97585-50ul
This item is not returnable.
View return policy
Description
AKR1B1 Polyclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| AKR1B1 | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1-5 μL/mg of lysate | |
| ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
| Produced in rabbits immunized with E. coli-derived Human AKR1B1 fragment. | |
| 50 μg | |
| Stem Cell Markers | |
| 231 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction