missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529718
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Rat (85%), Dog (91%), Pig (86%), Rabbit (89%), Rhesus Monkey (98%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.
Specifications
| AKR1B1 | |
| Polyclonal | |
| Unconjugated | |
| AKR1B1 | |
| ADR; Ahr1; Ahr-1; Akr1b1; Akr1b3; Akr1b4; Aldehyde reductase; aldehyde reductase 1; Aldehyde reductase 1 (low Km aldose reductase) (5.8 kb PstI fragment, probably the functional gene); aldo-keto reductase family 1; aldo-keto reductase family 1 member B; aldo-keto reductase family 1 member B1; aldo-keto reductase family 1 member B1; aldose reductase; Aldo-keto reductase family 1 member B3; aldo-keto reductase family 1, member B1 (aldose reductase); aldo-keto reductase family 1, member B3 (aldose reductase); aldo-keto reductase family 1, member B4 (aldose reductase); Aldor1; Aldose reductase; ALDR1; ALDRED; Alr; Alr2; ALR-P-I; AR; Lii5-2 CTCL tumor antigen; low Km aldose reductase; member B1 (aldose reductase); RATALDRED | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 231, 24192, 396816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.06 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P07943, P15121, P80276 | |
| AKR1B1 | |
| Recombinant protein encompassing a sequence within the center region of human AKR1B1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction