missing translation for 'onlineSavingsMsg'
Learn More

AKR1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15439-1-AP-150UL

 View more versions of this product

Product Code. 16824614

  • € 410.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.
Specifications

Specifications

AKR1B1
Polyclonal
Unconjugated
AKR1B1
ADR, AKR1B1, Aldehyde reductase, Aldose reductase, ALDR1, ALR2, AR
Rabbit
Antigen Affinity Chromatography
RUO
11677, 231, 24192
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07943, P15121, P45376
AKR1B1
AKR1B1 Fusion Protein Ag7142
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.