missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15439-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.Specifications
| AKR1B1 | |
| Polyclonal | |
| Unconjugated | |
| AKR1B1 | |
| ADR, AKR1B1, Aldehyde reductase, Aldose reductase, ALDR1, ALR2, AR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11677, 231, 24192 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07943, P15121, P45376 | |
| AKR1B1 | |
| AKR1B1 Fusion Protein Ag7142 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction