missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A3 Antibody (2E11), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97944JF646
This item is not returnable.
View return policy
Description
AKR7A3 Monoclonal antibody specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| AKR7A3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2E11 | |
| Western Blot, ELISA | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
| Recombinant human AKR7A3 (1-331aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 22977 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction