missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92684-0.1ml
This item is not returnable.
View return policy
Description
AKR7A3 Polyclonal antibody specifically detects AKR7A3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| AKR7A3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human AKR7A3 (NP_036199.2). MSRQLSRARPATVLGAMEMGRRMDAPTSAAVTRAFLERGHTEIDTAFVYSEGQSETILGGLGLRLGGSDCRVKIDTKAIPLFGNSLKPDSLRFQLETSLKRLQCPRVDLFYLHMPDHSTPVEETLRACHQLHQEGKFVELGLSNYAAWEVAEICTLCKSNGWILPTVYQGMYNAITRQVETELFPCLRHFGLRFYAFNPLAGGLLTGKYKYEDKNGKQPVGRFFGNTWAEMYRNRYWKEHHFEGIALVEKALQAAYGASAPSMTSATLRWMYHHSQLQGAHGDAVILGMSSLEQLEQNLAAAEEGPLEPAVVDAFNQAWHLVTHECPNYFR | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 22977 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction