Learn More
AKT [p Ser473] Antibody (545007) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB887JF635
This item is not returnable.
View return policy
Description
AKT Monoclonal antibody specifically detects AKT in Human,Mouse samples. It is validated for Western Blot,Simple Western
Specifications
| AKT | |
| Monoclonal | |
| Janelia Fluor 635 | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC | |
| Phosphopeptide containing the human Akt (S473) site | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot, Simple Western | |
| 545007 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cellular Signaling, Neuroscience | |
| 207 | |
| Store at 4°C in the dark. | |
| IgG1 |