missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1/2/3 Antibody (JE75-09), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32017
This item is not returnable.
View return policy
Description
AKT1/2/3 Monoclonal antibody specifically detects AKT1/2/3 in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| AKT1/2/3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1 | |
| Recombinant protein within Human AKT1/2/3 aa 231-480/ 480. (Uniprot: P31749) | |
| 100 μL | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Signal Transduction | |
| 207 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| JE75-09 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction