missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Antibody (V7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50101AF405
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| AKT1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50 mM Sodium Borate with 0.05% Sodium Azide | |
| AKT1 | |
| This AKT1 antibody was made to an ovalbumin-conjugated synthetic peptide KDPMQRLGGG (C-terminal sequence) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| V7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| 55.7 kDa | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction | |
| 207 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction