missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806076
This item is not returnable.
View return policy
Description
The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.Specifications
| AKT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P31749 | |
| AKT1 | |
| Synthetic peptide around the E17K mutation region of the human AKT1 conjugated to KLH | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B2 | |
| Unconjugated | |
| AKT1 | |
| AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 207 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction