missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT1, p Ser473 Antibody (104A282), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56749AF594
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKT1 (p Ser473) | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM Sodium Borate with 0.05% Sodium Azide | |
| AKT1 | |
| This AKT1 phospho Ser473 monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 473 of human AKT1. | |
| 0.1 mL | |
| 207 | |
| Human, Mouse, Rat, Rabbit | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 104A282 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| 55.7 kDa | |
| Primary | |
| Clone 104A282 detects specifically the Ser473 phosphorylated form of AKT1. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction