missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Akt1/PKBα Protein, Unactive

Product Code. p-7228033
Encompass_Preferred
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 17360114

Brand: Eurofins DiscoverX 14279

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unactive, N-terminal His6-tagged recombinant full-length human Akt1, for use in Kinase Assays.

Akt1 is an oncogenic AGC kinase that plays a critical role in regulating many processes including cell survival, metabolism, proliferation, growth and angiogenesis. AKT activation relies on the PI3K pathway, and is recognized as a critical node in the pathway. The E17 hotspot is the most characterized of AKT1 mutations, and has been shown to result in activation of the protein. Mutations in AKT1 have also been shown to confer resistance to allosteric kinase inhibitors in vitro.

GenBank M63167. The recombinant protein contains the amino acid substitution S478G with respect to this accession number. This conflict is reported inEMBL BE 206796. The residue coordinates in the native sequence are given.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 50% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol.
Molecular Weight (g/mol) 59 kDa
Quantity 50 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥65%
Protein Akt1/PKBa, unactive
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.