missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2 Antibody (302501) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB23152JF585
This item is not returnable.
View return policy
Description
AKT2 Monoclonal antibody specifically detects AKT2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| AKT2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt2, Asn2-Glu481, Accession # P31751 | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Signal Transduction | |
| 208 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 302501 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction