missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKT2 Monoclonal Antibody (4H7)
Mouse Monoclonal Antibody
€ 224.00 - € 558.00
Specifications
| Antigen | AKT2 |
|---|---|
| Clone | 4H7 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
12827762
|
Invitrogen™
MA1034 |
100 μg |
€ 558.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12837762
|
Invitrogen™
MA1034X |
20 μL |
€ 224.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKT2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ChIP
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone. Akt is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.Specifications
| AKT2 | |
| 1 mg/mL | |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
| P31751, P47197, Q60823 | |
| 11652, 208, 25233 | |
| Full length human recombinant protein of human AKT2 (NP_001617) produced in HEK293T cell. | |
| Antibody |
| 4H7 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat, Monkey | |
| Akt2 | |
| 2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 | |
| Akt2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title