Learn More
Invitrogen™ AKT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585321
Description
Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (85%), Xenopus laevis (83%), Dog (97%), Pig (98%), Rhesus Monkey (99%), Bovine (98%). Positive Control: HeLa, HA-AKT transfected HeLa cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone. Akt is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.
Specifications
| AKT2 | |
| Polyclonal | |
| Unconjugated | |
| Akt2 | |
| 2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 208 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.65 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P31751 | |
| Akt2 | |
| Recombinant protein encompassing a sequence within the center region of human Akt2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.