missing translation for 'onlineSavingsMsg'
Learn More

AKTIP Polyclonal Antibody
GREENER_CHOICE

Product Code. 15613495
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15613495 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613495 Supplier Invitrogen Supplier No. PA549195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AKTIP Polyclonal Antibody

AKTIP Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen AKTIP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene AKTIP
Gene Accession No. Q9H8T0
Gene Alias AKT interacting protein; AKT-interacting protein; AKTIP; AL023020; Fif; Ft; FT1; FTS; fused toes homolog; Fused toes protein; Fused toes protein homolog; thymoma viral proto-oncogene 1 interacting protein
Gene Symbols AKTIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 262-288 amino acids from the C-terminal region of human AKTIP.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.