missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aladin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546705
This item is not returnable.
View return policy
Description
Peptide sequence: EHNNELVTGS SYESPPPDFR GQWINLPVLQ LTKDPLKTPG RLDHGTRTAF.
The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Aladin | |
| Polyclonal | |
| Unconjugated | |
| AAAS | |
| AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRG9 | |
| AAAS | |
| Synthetic peptide directed towards the N-terminal of human AAAS (aa 20-69). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction