missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alanyl tRNA synthetase Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97723MFV450
This item is not returnable.
View return policy
Description
Alanyl tRNA synthetase Polyclonal antibody specifically detects Alanyl tRNA synthetase in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Alanyl tRNA synthetase | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| alanine tRNA ligase 1, cytoplasmic, Alanine--tRNA ligase, alanyl-tRNA synthetase, alanyl-tRNA synthetase, cytoplasmic, AlaRS, CMT2N, EC 6.1.1, EC 6.1.1.7, Renal carcinoma antigen NY-REN-42 | |
| Produced in rabbits immunized with E. coli-derived Human Alanyl tRNA synthetase fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 16 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction