Learn More
ALAS1 Mouse, anti-Human, Clone: 1B1E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68567-1-IG-150UL
This item is not returnable.
View return policy
Description
The ALAS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALAS1. This antibody recognizes human, rat, pig antigen. The ALAS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ALAS1 | |
| 1B1E4 | |
| ALAS, ALAS H, ALAS1, ALAS3, ALASH, Delta ALA synthase 1, MIG4 | |
| 150 μL | |
| 211 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Rat | |
| 150 kDa | |
| IgG1 |