Learn More
ALAS1 Mouse, anti-Human, Clone: 1B1E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68567-1-IG-150UL
 View more versions of this product
Product Code. 18094995
  • € 410.00 / 150µL
Description
The ALAS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALAS1. This antibody recognizes human, rat, pig antigen. The ALAS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ALAS1
1B1E4
ALAS, ALAS H, ALAS1, ALAS3, ALASH, Delta ALA synthase 1, MIG4
150 μL
211
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Pig, Rat
150 kDa