missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALAS1 Recombinant Rabbit Monoclonal Antibody (23GB3190), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | ALAS1 |
|---|---|
| Clone | 23GB3190 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088195
![]() |
Invitrogen™
MA553358 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088879
|
Invitrogen™
MA553359 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALAS1 Recombinant Monoclonal antibody specifically detects ALAS1 in Human, Rat samples. It is validated for Western BlotSpecifications
| ALAS1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| P13195, P13196 | |
| 211, 65155 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3190 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5'-aminolevulinate synthase 1; 5-aminolevulinate synthase 1; 5-aminolevulinate synthase, nonspecific, mitochondrial; 5-aminolevulinic acid synthase 1; ALAS; ALA-S; ALAS1; Alas-1; ALAS3; ALASH; Alas-h; ALAS-N; aminolevulinate synthase H; aminolevulinate, delta-, synthase 1; aminolevulinic acid synthase 1; delta-ALA synthase 1; delta-ALA synthetase; delta-aminolevulinate synthase 1; MIG4; migration-inducing protein 4; OK/SW-cl.121; succinyl-CoA: glycine C-succinyl transferase | |
| A synthetic peptide of human Alas1 (600-640AA). | |
| Primary | |
| -20°C | |
| ALAS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title