Learn More
Emma Safety Footwear Alaska XD Safety Shoes
Product Code. 15848787
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15848787
Brand: Emma Safety Footwear MM30356640
Only null left
Add to basket
Add to basket
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Alaska XD
Material Oiled Nubuck
Size 40