missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Albendazole Monoclonal Antibody (3H2)
GREENER_CHOICE

Product Code. 17979922
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979922 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17979922 Supplier Invitrogen™ Supplier No. MA549633

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Anthelmintics or anti-helminthics are a class of drugs that are effective against a range of intestinal parasitic worms (helminths). Parasitic helminths must maintain an appropriate feeding site. Nematodes and trematodes must actively ingest and move food through their digestive tracts to maintain an appropriate energy state; these together with reproductive processes require a well defined and developed neuromuscular coordination. Anthelmintic treatment is a multi-targeting system designed to interfere with the integrity of parasite cells. The pharmacologic basis of the treatment for helminths involves the targeting of neuromuscular coordination, or protective mechanisms against host immunity, which lead to starvation, paralysis, and expulsion of the parasite. The benzimidazole class of drugs were introduced in 1961 and interfere with the parasite's ATP pathway on a cellular level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Albendazole
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3H2
Concentration 6.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.6
Host Species Mouse
Immunogen Conjugate of ABZ hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.