Learn More
Albuterol-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Albuterol-d4 |
| CAS | 2517375-34-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13H17D4NO3 |
| Quantity | 1 mg |
| Synonym | α1-[[(1,1-Dimethylethyl)amino]methyl]-4-hydroxy-1,3-benzenedimethanol-d4, Salbutamol-d4 |
| InChI Formula | InChI=1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3/i7D,8D2,12D |
| SMILES | OC1=CC=C(C(C([2H])NC(C)(C)C)([2H])O)C=C1C([2H])([2H])O |
| IUPAC Name | 4-[2-(tert-butylamino)-1,2-dideuterio-1-hydroxyethyl]-2-[dideuterio(hydroxy)methyl]phenol |
| Molecular Weight (g/mol) | 243.34 |
| Show More |