missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALCAM/CD166 Antibody (008), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90954PE
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Cynomolgus Monkey samples. It is validated for Western Blot
Specifications
| ALCAM/CD166 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
| Western Blot | |
| 008 | |
| Western Blot | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus ALCAM/CD166 (Accession#: G7NZQ8; Met1-Ala526). | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 214 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction