missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALCAM/CD166 Antibody (105902), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6561M
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
Specifications
| ALCAM/CD166 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 105902 | |
| Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166, Trp28-Ala526, Accession # Q13740 | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 214 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction