missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 1B Antibody (OTI4F12), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70092APC
This item is not returnable.
View return policy
Description
alcohol dehydrogenase 1B Monoclonal specifically detects alcohol dehydrogenase 1B in Human, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| alcohol dehydrogenase 1B | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| ADH1B | |
| Full length recombinant protein of human alcohol dehydrogenase 1B (NP_000659) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine | |
| IgG2a |
| Flow Cytometry | |
| OTI4F12 | |
| Flow Cytometry | |
| ADH, beta subunit, ADH2alcohol dehydrogenase 1B, alcohol dehydrogenase 1B (class I), beta polypeptide, alcohol dehydrogenase 2 (class I), beta polypeptide, Alcohol dehydrogenase subunit beta, aldehyde reductase, DKFZp686C06125, EC 1.1.1, EC 1.1.1.1 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 125.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction