Learn More
alcohol dehydrogenase 1B Antibody (OTI4F12), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-70092PECY7
Product Code. 18031596
  • € 804.00 / 0.10mL
Description
alcohol dehydrogenase 1B Monoclonal specifically detects alcohol dehydrogenase 1B in Human, Rat, Canine samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alcohol dehydrogenase 1B
Monoclonal
PE-Cyanine7
PBS with 0.05% Sodium Azide
ADH1B
Full length recombinant protein of human alcohol dehydrogenase 1B (NP_000659) produced in HEK293T cells.
0.1 mL
Primary
Human, Rat, Canine
IgG2a
Flow Cytometry
OTI4F12
Flow Cytometry
ADH, beta subunit, ADH2alcohol dehydrogenase 1B, alcohol dehydrogenase 1B (class I), beta polypeptide, alcohol dehydrogenase 2 (class I), beta polypeptide, Alcohol dehydrogenase subunit beta, aldehyde reductase, DKFZp686C06125, EC 1.1.1, EC 1.1.1.1
Mouse
Affinity Purified
Cancer
125.0
Store at 4°C in the dark. Do not freeze.