missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK6 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76877JF646
This item is not returnable.
View return policy
Description
SLITRK6 Polyclonal antibody specifically detects SLITRK6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLITRK6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human Slitrk6. The immunogen is located within amino acids 250 - 300 of Slitrk6. Amino Acid Squence: TTPVYEEHEDPSG | |
| 0.1 mL | |
| Neuroscience | |
| 84189 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction